4-Methoxy-2-nitrophenylboronsäure

Lieferant: Apollo Scientific

Synonyme: (4-Methoxy-2-nitrophenyl)boronic acid

OR59360-250MG
APOSOR59360-250MGEA 277 EUR
APOSOR59360-250MG
4-Methoxy-2-nitrophenylboronsäure
4-Methoxy-2-nitrophenylboronsäure
Formel: C₇H₈BNO₅
Molecular Weight: 196,950 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09878340
CAS-Nummer: 860034-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR