4-Brom-6-chlor-3-pyridinboronsäure

Lieferant: Apollo Scientific

Synonyme: 4-Brom-6-chlor-3-pyridinylboronsäure, 4-Brom-6-chlorpyridin-3-boronsäure

OR59363-250MG
APOSOR59363-250MGEA 288 EUR
APOSOR59363-250MG
4-Brom-6-chlor-3-pyridinboronsäure
4-Bromo-6-chloro-3-pyridineboronic acid
Formel: C₅H₄BBrClNO₂
Molecular Weight: 236,26 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09878338
CAS-Nummer: 957062-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR