3-Chlor-N,2-dimethylanilin Hydrochlorid

Lieferant: Apollo Scientific

OR59371-1G
APOSOR59371-1GEA 176 EUR
APOSOR59371-1G
3-Chlor-N,2-dimethylanilin Hydrochlorid
3-Chlor-N,2-dimethylanilin Hydrochlorid
Formel: C₈H₁₁Cl₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD09878314
CAS-Nummer: 957062-82-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR