2-Brom-5-chlor-4-methylanilin

Lieferant: Apollo Scientific

OR59373-1G
APOSOR59373-1GEA 30.6 EUR
APOSOR59373-1G
2-Brom-5-chlor-4-methylanilin
2-Brom-5-chlor-4-methylanilin
Formel: C₇H₇BrClN
Molecular Weight: 220,4942 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00806533
CAS-Nummer: 102170-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR