(±)-Indolin-2-carbonsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: Indoline-2-carboxylic acid

OR59374-100G OR59374-25G OR59374-5G
APOSOR59374-100GEA 132 EUR
APOSOR59374-100G APOSOR59374-25G APOSOR59374-5G
(±)-Indolin-2-carbonsäure
(±)-Indolin-2-carbonsäure
Formel: C₉H₉NO₂
Molecular Weight: 163.18 g/mol
Schmelzpunkt: 168 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010635
CAS-Nummer: 78348-24-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR