4-Bromo-1-[(2-bromophenyl)sulphonyl]-1H-pyrazole

Lieferant: Apollo Scientific

OR59377-1G
APOSOR59377-1GEA 176 EUR
APOSOR59377-1G
4-Bromo-1-[(2-bromophenyl)sulphonyl]-1H-pyrazole
4-Bromo-1-[(2-bromophenyl)sulphonyl]-1H-pyrazole
Formel: C₉H₆Br₂N₂O₂S
Molecular Weight: 366,029 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878370
CAS-Nummer: 957062-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR