6-Bromo-2-cyclohexylquinazolin-4(3H)-one

Lieferant: Apollo Scientific

OR59392-1G
APOSOR59392-1GEA 212 EUR
APOSOR59392-1G
6-Bromo-2-cyclohexylquinazolin-4(3H)-one
6-Bromo-2-cyclohexylquinazolin-4(3H)-one
Formel: C₁₄H₁₅BrN₂O
Molecular Weight: 307,186 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878379
CAS-Nummer: 1000339-29-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR