(2-Bromophenyl)(dimethylamino)acetonitrile

Lieferant: Apollo Scientific
Achtung

OR59401-1G
APOSOR59401-1GEA 176 EUR
APOSOR59401-1G
(2-Bromophenyl)(dimethylamino)acetonitrile
(2-Bromophenyl)(dimethylamino)acetonitrile
Formel: C₁₀H₁₁BrN₂
Molecular Weight: 239,112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878365
CAS-Nummer: 299215-38-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR