Methyl-4-methoxynicotinat 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-Methoxynicotinate

OR59403-250MG
APOSOR59403-250MGEA 21.9 EUR
APOSOR59403-250MG
Methyl-4-methoxynicotinat 95%
Methyl-4-methoxynicotinat
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
Schmelzpunkt: 82…85 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09864940
CAS-Nummer: 10177-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR