5-Bromo-2-[(4-methoxybenzyl)oxy]pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-((4-methoxybenzyl)oxy)pyridine, 4-{[(5-Bromopyridin-2-yl)oxy]methyl}anisole, 1-{[(5-Bromopyridin-2-yl)oxy]methyl}-4-methoxybenzene

OR59411-1G
APOSOR59411-1GEA 176 EUR
APOSOR59411-1G
5-Bromo-2-[(4-methoxybenzyl)oxy]pyridine
5-Bromo-2-[(4-methoxybenzyl)oxy]pyridine
Formel: C₁₃H₁₂BrNO₂
Molecular Weight: 294,144 g/mol
Schmelzpunkt: 76…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689424
CAS-Nummer: 663955-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR