4-[(3-Chloro-2-methylphenyl)sulphamoyl]benzeneboronic acid

Lieferant: Apollo Scientific

Synonyme: (4-(N-(3-Chloro-2-methylphenyl)sulphamoyl)phenyl)boronic acid

OR59412-1G
APOSOR59412-1GEA 306 EUR
APOSOR59412-1G
4-[(3-Chloro-2-methylphenyl)sulphamoyl]benzeneboronic acid
4-[(3-Chloro-2-methylphenyl)sulphamoyl]benzeneboronic acid
Formel: C₁₃H₁₃BClNO₄S
Molecular Weight: 325,576 g/mol
Schmelzpunkt: 189…191 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09878330
CAS-Nummer: 957066-10-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR