4-[3-(2-Acetylhydrazino)-3-oxopropyl]benzeneboronic acid

Lieferant: Apollo Scientific

OR59415-1G
APOSOR59415-1GEA 205 EUR
APOSOR59415-1G
4-[3-(2-Acetylhydrazino)-3-oxopropyl]benzeneboronic acid
4-[3-(2-Acetylhydrazino)-3-oxopropyl]benzeneboronic acid
Formel: C₁₁H₁₅BN₂O₄
Molecular Weight: 250,059 g/mol
Schmelzpunkt: 218…221 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09878348
CAS-Nummer: 957066-08-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR