2-Chlor-4-methoxy-5-(methoxycarbonyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

OR59416-500MG
APOSOR59416-500MGEA 328 EUR
APOSOR59416-500MG
2-Chlor-4-methoxy-5-(methoxycarbonyl)phenylboronsäure
2-Chlor-4-methoxy-5-(methoxycarbonyl)phenylboronsäure
Formel: C₉H₁₀BClO₅
Schmelzpunkt: 135…137 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09878324
CAS-Nummer: 957066-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR