(S)-1-Boc-2-methylpiperazin

Lieferant: Apollo Scientific

Synonyme: (S)-2-Methyl-1-piperazinecarboxylic acid tert-Butyl ester, tert-Butyl (S)-2-Methyl-1-piperazinecarboxylate, (S)-tert-Butyl 2-methylpiperazine-1-carboxylate, (S)-1-Boc-2-methylpiperazine, (S)-1-(tert-Butoxycarbonyl)-2-methylpiperazine

OR59442-1G
APOSOR59442-1GEA 21.9 EUR
APOSOR59442-1G
(S)-1-Boc-2-methylpiperazin
(S)-1-Boc-2-methylpiperazin
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,2780 g/mol
MDL: MFCD01862121
CAS-Nummer: 169447-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR