5-Brom-o-toluylsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-methylbenzoic acid

OR59443-100G
APOSOR59443-100GEA 116 EUR
APOSOR59443-100G
5-Brom-o-toluylsäure
5-Brom-o-toluylsäure
Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
Schmelzpunkt: 167…171 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00267350
CAS-Nummer: 79669-49-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR