4-Nitro-o-toluylsäure

Lieferant: Apollo Scientific
Achtung

OR59451-5G OR59451-25G OR59451-100G
APOSOR59451-5GEA 21.9 EUR
APOSOR59451-5G APOSOR59451-25G APOSOR59451-100G
4-Nitro-o-toluylsäure
4-Nitro-o-toluylsäure
Formel: C₈H₇NO₄
Molecular Weight: 181.15 g/mol
Schmelzpunkt: 150…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00210697
CAS-Nummer: 1975-51-5
EINECS: 217-828-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR