1-(3-Aminopyridin-4-yl)ethanone

Lieferant: Apollo Scientific

OR59461-1G
APOSOR59461-1GEA 85.7 EUR
APOSOR59461-1G
1-(3-Aminopyridin-4-yl)ethanone
1-(3-Aminopyridin-4-yl)ethanone
Formel: C₇H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD07375078
CAS-Nummer: 13210-52-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR