6-Chlor-2-pyridinboronsäurepinakolester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Chloropyridine-2-boronic acid pinacol ester

OR59467-250MG
APOSOR59467-250MGEA 21.9 EUR
APOSOR59467-250MG
6-Chlor-2-pyridinboronsäurepinakolester 98%
6-Chlor-2-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BClNO₂
Molecular Weight: 239.51 g/mol
Schmelzpunkt: 109…111 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD06798259
CAS-Nummer: 652148-92-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR