Methyl-2-(3-aminophenyl)acetate hydrochloride

Lieferant: Apollo Scientific
Achtung

OR59469-1G OR59469-5G OR59469-25G
APOSOR59469-1GEA 0 EUR
APOSOR59469-1G APOSOR59469-5G APOSOR59469-25G
Methyl-2-(3-aminophenyl)acetate hydrochloride
Methyl-2-(3-aminophenyl)acetate hydrochloride
Formel: C₉H₁₂ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11501071
CAS-Nummer: 150319-83-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR