5-Cyanouracil

Lieferant: Apollo Scientific
Gefahr

Synonyme: Uracil-5-carbonitrile, 2,4-Dihydroxypyrimidine-5-carbonitrile, 2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile, 5-Cyano-2,4-dioxo-1,2,3,4-tetrahydropyrimidine

OR5947-1G
APOSOR5947-1GEA 42.2 EUR
APOSOR5947-1G
5-Cyanouracil
5-Cyanouracil
Formel: C₅H₃N₃O₂
Molecular Weight: 137,10 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00128255
CAS-Nummer: 4425-56-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR