4-Methoxypiperidin
Lieferant: Apollo Scientific
|
Achtung
|
OR59470-1G
APOSOR59470-1GEA
21.9
EUR
APOSOR59470-1G
4-Methoxypiperidin
4-Methoxypiperidin
Formel:
C₆H₁₄ClNO Molecular Weight: 115,1700 g/mol Siedepunkt: 103 °C/113mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115010 CAS-Nummer: 4045-24-3 EINECS: 223-742-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...