2-Amino-5-bromo-3-(hydroxymethyl)pyridine hydrobromide

Lieferant: Apollo Scientific
Achtung

OR59471-1G OR59471-250MG OR59471-500MG
APOSOR59471-1GEA 0 EUR
APOSOR59471-1G APOSOR59471-250MG APOSOR59471-500MG
2-Amino-5-bromo-3-(hydroxymethyl)pyridine hydrobromide
2-Amino-5-bromo-3-(hydroxymethyl)pyridine hydrobromide
Formel: C₆H₈Br₂N₂O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 443956-55-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR