2-Amino-5-bromo-3-(hydroxymethyl)pyridine hydrobromide
Lieferant: Apollo Scientific
|
Achtung
|
OR59471-1G
OR59471-250MG
OR59471-500MG
APOSOR59471-1GEA
0
EUR
APOSOR59471-1G
APOSOR59471-250MG
APOSOR59471-500MG
2-Amino-5-bromo-3-(hydroxymethyl)pyridine hydrobromide
2-Amino-5-bromo-3-(hydroxymethyl)pyridine hydrobromide
Formel:
C₆H₈Br₂N₂O Lagertemperatur: Raumtemperatur |
CAS-Nummer:
443956-55-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...