Ethyl thiomorpholine-3-carboxylate hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Ethoxycarbonyl)thiomorpholine hydrochloride, Ethyl 1,4-thiazinane-3-carboxylate hydrochloride

OR59472-1G
APOSOR59472-1GEA 318 EUR
APOSOR59472-1G
Ethyl thiomorpholine-3-carboxylate hydrochloride
Ethyl thiomorpholine-3-carboxylate hydrochloride
Formel: C₇H₁₃NO₂S·ClH
Molecular Weight: 211,71 g/mol
Schmelzpunkt: 133…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00107968
CAS-Nummer: 159381-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR