2-Amino-p-kresol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR59481-25G
OR59481-100G
APOSOR59481-25GEA
21.9
EUR
APOSOR59481-25G
APOSOR59481-100G
2-Amino-p-kresol 97%
2-Amino-p-kresol
Formel:
C₇H₉NO Schmelzpunkt: 133…138 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007699 CAS-Nummer: 95-84-1 EINECS: 202-457-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...