5-Methyl-thiophene-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR5951-1G OR5951-250MG OR5951-5G
APOSOR5951-1GEA 52.2 EUR
APOSOR5951-1G APOSOR5951-250MG APOSOR5951-5G
5-Methyl-thiophene-3-carboxylic acid
5-Methyl-thiophene-3-carboxylic acid
Formel: C₆H₆O₂S
Lagertemperatur: Kühlschrank
MDL: MFCD01680229
CAS-Nummer: 19156-50-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR