4'-Brom-3-chlorpropiophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4'-Brom-β-chlorpropiophenon
OR5955-100G
APOSOR5955-100GEA
177
EUR
APOSOR5955-100G
4'-Brom-3-chlorpropiophenon 98%
4'-Brom-3-chlorpropiophenon
Formel:
C₉H₈BrClO Molecular Weight: 247.52 g/mol Siedepunkt: 301 Schmelzpunkt: 54…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00018979 CAS-Nummer: 31736-73-9 EINECS: 250-784-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...