4-(4-(2-Hydroxyethyl)-1-piperazinylcarbonyl)phenylboronsäure Hydrochlorid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-(4-(2-Hydroxyethyl)piperazine-1-carbonyl)phenyl)boronic acid hydrochloride

OR5961-1G
APOSOR5961-1GEA 183 EUR
APOSOR5961-1G
4-(4-(2-Hydroxyethyl)-1-piperazinylcarbonyl)phenylboronsäure Hydrochlorid 97%
4-(4-(2-Hydroxyethyl)-1-piperazinylcarbonyl)phenylboronsäure Hydrochlorid
Formel: C₁₃H₁₉BN₂O₄·HCl
Molecular Weight: 314,57 g/mol
Schmelzpunkt: 100…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027218
CAS-Nummer: 913835-44-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR