4-(4-Methylpiperazine-1-carbonyl)phenylboronsäure Hydrochlorid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid hydrochloride

OR5965-1G
APOSOR5965-1GEA 270 EUR
APOSOR5965-1G
4-(4-Methylpiperazine-1-carbonyl)phenylboronsäure Hydrochlorid 95%
4-(4-Methylpiperazine-1-carbonyl)phenylboronsäure Hydrochlorid
Formel: C₁₂H₁₈BClN₂O₃
Molecular Weight: 284.55 g/mol
Schmelzpunkt: 187…189 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09027217
CAS-Nummer: 913835-43-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR