4-[2-(Diethylamino)ethylcarbamoyl]benzeneboronic acid hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

OR5966-1G
APOSOR5966-1GEA 176 EUR
APOSOR5966-1G
4-[2-(Diethylamino)ethylcarbamoyl]benzeneboronic acid hydrochloride 98%
4-[2-(Diethylamino)ethylcarbamoyl]benzeneboronic acid hydrochloride
Formel: C₁₃H₂₁BN₂O₃·HCl
Molecular Weight: 300,59 g/mol
Schmelzpunkt: 65…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027220
CAS-Nummer: 913835-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR