4-(Piperidin-1-carbonyloxy)phenylboronsäurepinakolester 98%

Lieferant: Apollo Scientific

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl piperidine-1-carboxylate

OR5967-1G
APOSOR5967-1GEA 250 EUR
APOSOR5967-1G
4-(Piperidin-1-carbonyloxy)phenylboronsäurepinakolester 98%
4-(Piperidin-1-carbonyloxy)phenylboronsäurepinakolester
Formel: C₁₈H₂₆BNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09027287
CAS-Nummer: 913836-28-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR