4-[(Cyclopentylcarbamoyl)amino]benzeneboronic acid, pinacol ester 95%

Lieferant: Apollo Scientific

Synonyme: 4-(3-Cyclopentylureido)benzeneboronic acid, pinacol ester

OR5968-1G
APOSOR5968-1GEA 176 EUR
APOSOR5968-1G
4-[(Cyclopentylcarbamoyl)amino]benzeneboronic acid, pinacol ester 95%
4-[(Cyclopentylcarbamoyl)amino]benzeneboronic acid, pinacol ester
Formel: C₁₈H₂₇BN₂O₃
Molecular Weight: 330,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027290
CAS-Nummer: 874297-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR