4-{[(Furan-2-ylmethyl)carbamoyl]amino}benzeneboronic acid, pinacol ester 98%

Lieferant: Apollo Scientific

OR5969-1G
APOSOR5969-1GEA 176 EUR
APOSOR5969-1G
4-{[(Furan-2-ylmethyl)carbamoyl]amino}benzeneboronic acid, pinacol ester 98%
4-{[(Furan-2-ylmethyl)carbamoyl]amino}benzeneboronic acid, pinacol ester
Formel: C₁₈H₂₃BN₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09027292
CAS-Nummer: 874297-85-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR