tert-Butyl-3-(benzyloxy)-4-brombenzoat 98%

Lieferant: Apollo Scientific

OR5970-1G
APOSOR5970-1GEA 176 EUR
APOSOR5970-1G
tert-Butyl-3-(benzyloxy)-4-brombenzoat 98%
tert-Butyl-3-(benzyloxy)-4-brombenzoat
Formel: C₁₈H₁₉BrO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09027266
CAS-Nummer: 247186-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR