4-[(Propylcarbamoyl)amino]benzeneboronic acid, pinacol ester 98%

Lieferant: Apollo Scientific

Synonyme: 1-Propyl-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea, 4-(3-Propylureido)benzeneboronic acid, pinacol ester

OR5972-1G
APOSOR5972-1GEA 159 EUR
APOSOR5972-1G
4-[(Propylcarbamoyl)amino]benzeneboronic acid, pinacol ester 98%
4-[(Propylcarbamoyl)amino]benzeneboronic acid, pinacol ester
Formel: C₁₆H₂₅BN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09027288
CAS-Nummer: 874291-01-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR