6-(1H-Imidazol-1-yl)nicotinonitrile
Lieferant: Apollo Scientific
OR5977-1G
APOSOR5977-1GEA
129
EUR
APOSOR5977-1G
6-(1H-Imidazol-1-yl)nicotinonitrile
6-(1H-Imidazol-1-yl)nicotinonitrile
Formel:
C₉H₆N₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08691169 CAS-Nummer: 923156-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...