3-(1H-Imidazol-1-yl)pyrazine-2-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR5978-1G
APOSOR5978-1GEA
163
EUR
APOSOR5978-1G
3-(1H-Imidazol-1-yl)pyrazine-2-carbonitrile
3-(1H-Imidazol-1-yl)pyrazine-2-carbonitrile
Formel:
C₈H₅N₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09414743 CAS-Nummer: 342412-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...