4-Piperazin-1-ylbenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Piperazin-1-yl)benzoic acid
OR59793-1G
APOSOR59793-1GEA
347
EUR
APOSOR59793-1G
4-Piperazin-1-ylbenzoic acid
4-Piperazin-1-ylbenzoic acid
Formel:
C₁₁H₁₄N₂O₂ Molecular Weight: 206,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03444417 CAS-Nummer: 85474-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...