2,5-Dibromisonicotinsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,5-Dibromopyridine-4-carboxylic acid

OR59812-5G
APOSOR59812-5GEA 74.1 EUR
APOSOR59812-5G
2,5-Dibromisonicotinsäure
2,5-Dibromisonicotinsäure
Formel: C₆H₃Br₂NO₂
Molecular Weight: 280.9000 g/mol
Schmelzpunkt: 174…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08741485
CAS-Nummer: 942473-59-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR