2,3-Dibromisonicotinsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dibromopyridine-4-carboxylic acid

OR59813-1G
APOSOR59813-1GEA 136 EUR
APOSOR59813-1G
2,3-Dibromisonicotinsäure 97%
2,3-Dibromisonicotinsäure
Formel: C₆H₃Br₂NO₂
Molecular Weight: 280,90 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09997708
CAS-Nummer: 1020056-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR