α-Brom-3-nitrotoluol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
α-Brom-m-nitrotoluol, 3-Nitrobenzylbromid, m-Nitrobenzylbromid
OR59814-100G
APOSOR59814-100GEA
148
EUR
APOSOR59814-100G
α-Brom-3-nitrotoluol
α-Brom-3-nitrotoluol
Formel:
C₇H₆BrNO₂ Molecular Weight: 216,03 g/mol Schmelzpunkt: 58…59 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007271 CAS-Nummer: 3958-57-4 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...