2-Methylnicotinsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methyl-3-pyridincarbonsäure, 2-Methylpyridin-3-carbonsäure

OR59824-1G
APOSOR59824-1GEA 24.8 EUR
APOSOR59824-1G
2-Methylnicotinsäure
2-Methylnicotinsäure
Formel: C₇H₇NO₂
Molecular Weight: 137.14 g/mol
Schmelzpunkt: 228…230 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013449
CAS-Nummer: 3222-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR