3,4-Dihydroxypyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: Pyridine-3,4-diol

OR59831-1G
APOSOR59831-1GEA 173 EUR
APOSOR59831-1G
3,4-Dihydroxypyridine
3,4-Dihydroxypyridine
Formel: C₅H₅NO₂
Molecular Weight: 111,099 g/mol
Schmelzpunkt: 242…244 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01691806
CAS-Nummer: 10182-48-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR