2-Hydroxy-4-methylpyrimidine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Methylpyrimidin-2-ol hydrochloride

OR59839-25G
APOSOR59839-25GEA 21.9 EUR
APOSOR59839-25G
2-Hydroxy-4-methylpyrimidine hydrochloride
2-Hydroxy-4-methylpyrimidine hydrochloride
Formel: C₅H₇ClN₂O
Molecular Weight: 146,57 g/mol
Schmelzpunkt: 243 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012782
CAS-Nummer: 5348-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR