4-Hydrazinoquinoline hydrochloride

Lieferant: Apollo Scientific
Achtung

OR59842-1G OR59842-250MG OR59842-5G
APOSOR59842-1GEA 364 EUR
APOSOR59842-1G APOSOR59842-250MG APOSOR59842-5G
4-Hydrazinoquinoline hydrochloride
4-Hydrazinoquinoline hydrochloride
Formel: C₉H₁₀ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01861876
CAS-Nummer: 68500-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR