5-(Hydroxymethyl)-2-methyl-1,3-oxazole 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Methyl-1,3-oxazol-5-yl)methanol
OR59863-1G
APOSOR59863-1GEA
696
EUR
APOSOR59863-1G
5-(Hydroxymethyl)-2-methyl-1,3-oxazole 95%
5-(Hydroxymethyl)-2-methyl-1,3-oxazole
Formel:
C₅H₇NO₂ Molecular Weight: 113,115 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11101415 CAS-Nummer: 1065073-48-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...