Methyl-5-amino-2-furoat

Lieferant: Apollo Scientific

Synonyme: Methyl 5-aminofuran-2-carboxylate

OR59869-1G
APOSOR59869-1GEA 71.2 EUR
APOSOR59869-1G
Methyl-5-amino-2-furoat
Methyl-5-amino-2-furoat
Formel: C₆H₇NO₃
MDL: MFCD00051661
CAS-Nummer: 22600-30-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR