2-Amino-5-pyrimidinboronsäurepinakolester 98%

Lieferant: Apollo Scientific
Achtung

OR59880-10G OR59880-25G
APOSOR59880-10GEA 85.7 EUR
APOSOR59880-10G APOSOR59880-25G
2-Amino-5-pyrimidinboronsäurepinakolester 98%
2-Amino-5-pyrimidinboronsäurepinakolester
Formel: C₁₀H₁₆BN₃O₂
Schmelzpunkt: 209…211 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06795685
CAS-Nummer: 402960-38-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR