2-Brom-5-chlorphenol 97%

Lieferant: Apollo Scientific
Achtung

OR59899-5G
APOSOR59899-5GEA 27.7 EUR
APOSOR59899-5G
2-Brom-5-chlorphenol 97%
2-Brom-5-chlorphenol
Formel: C₆H₄BrClO
Molecular Weight: 207.45 g/mol
Schmelzpunkt: 63…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00142870
CAS-Nummer: 13659-23-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR