3-(Dimethylcarbamoyl)phenylboronsäurepinakolester 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N,N-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
OR59907-1G
OR59907-5G
APOSOR59907-1GEA
105
EUR
APOSOR59907-1G
APOSOR59907-5G
3-(Dimethylcarbamoyl)phenylboronsäurepinakolester 98%
3-(Dimethylcarbamoyl)phenylboronsäurepinakolester
Formel:
C₁₅H₂₂BNO₃ Schmelzpunkt: 90…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05863907 CAS-Nummer: 832114-07-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...